Learn More
DWK Life Sciences O-Ring, red, FEP coated, not suitable for desiccators
Seal is suitable for the tight sealing of flat flange vessels with annular groove.
Brand:  DWK Life Sciences 292226103
83.55 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.02200kg
 View more versions of this product
Product Code. 10613602
  • 87.95 EUR / Each
Estimated Shipment: 22-05-2024
Description
Flat flange vessels are large-volume reaction vessels consisting of a lower part and a lid. These two parts are held together by a clamping ring. The red O-ring made of silicone is seamlessly coated with FEP and serves as an optimal seal for flat flange vessels with an annular groove. It prevents jamming of the ground surfaces between the base and the lid and is convincing due to its good elasticity in combination with outstanding chemical resistance.
  • Accessories for flat flange vessels, comprising an elastic, silicone core with seamless FEP coating that encloses the ring
  • The combination of these high-quality materials achieves good elasticity in conjunction with outstanding chemical resistance
Specifications
Specifications
Tight closure of flange vessels
1 Ea.
Silicone, FEP
215 mm