Learn More
O-Phosphorylethanolamine, 98%, Thermo Scientific Chemicals
Catalog Number p-2689919
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
1071-23-4
C2H8NO4P
141.06
MFCD00008178
SUHOOTKUPISOBE-UHFFFAOYSA-N
o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate
1015
CHEBI:17553
2-aminoethyl dihydrogen phosphate
C(COP(=O)(O)O)N
1071-23-4
C2H8NO4P
141.06
MFCD00008178
SUHOOTKUPISOBE-UHFFFAOYSA-N
o-phosphoethanolamine, o-phosphorylethanolamine, phosphoethanolamine, o-phosphocolamine, colamine phosphate, phosphorylethanolamine, ethanolamine phosphate, phosphonoethanolamine, ethanolamine o-phosphate, mono 2-aminoethyl phosphate
1015
CHEBI:17553
2-aminoethyl dihydrogen phosphate
C(COP(=O)(O)O)N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 241.0°C to 243.0°C
Assay Percent Range 97.5 % min. (Titration with HClO4)
Packaging Glass Bottle
Linear Formula H2NCH2CH2OP(O)(OH)2
Quantity 5 g
Beilstein 04,IV,1415
Solubility Information Solubility in water: soluble.
Formula Weight 141.06
Percent Purity 98%
Chemical Name or Material O-Phosphorylethanolamine, 98%
compliance-icons
Product Identifier
  • O-Phosphorylethanolamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.