missing translation for 'onlineSavingsMsg'
Learn More

O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate, 98%, Thermo Scientific™

Catalog Number p-4228362
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
105379-24-6
C15H20N5O·F6P
431.32
XKTRAGMCMJYRRN-UHFFFAOYSA-N
hbpyu, benzotriazol-1-yloxy dipyrrolidinocarbenium hexafluorophosphate, 1h-benzo d 1,2,3 triazol-1-yl di pyrrolidin-1-yl methylene oxonium hexafluorophosphate v, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, hbpyu, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy benzotriazole hexafluorophosphate, 1-1,2,3-benzotriazol-1-yloxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, benzotriazol-1-yloxy dipyrrolidinocarbenium hexaf, 1,2,3-benzotriazol-1-yloxy bis pyrrolidin-1-yl methylium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy benzotriazole hexafluorophosphate
13685355
1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
This item is not returnable. View return policy
105379-24-6
C15H20N5O·F6P
431.32
XKTRAGMCMJYRRN-UHFFFAOYSA-N
hbpyu, benzotriazol-1-yloxy dipyrrolidinocarbenium hexafluorophosphate, 1h-benzo d 1,2,3 triazol-1-yl di pyrrolidin-1-yl methylene oxonium hexafluorophosphate v, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, hbpyu, o-benzotriazol-1-yl-n,n,n',n'-bis tetramethylene uronium hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy benzotriazole hexafluorophosphate, 1-1,2,3-benzotriazol-1-yloxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate, benzotriazol-1-yloxy dipyrrolidinocarbenium hexaf, 1,2,3-benzotriazol-1-yloxy bis pyrrolidin-1-yl methylium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy benzotriazole hexafluorophosphate
13685355
1-[pyrrolidin-1-ium-1-ylidene(pyrrolidin-1-yl)methoxy]benzotriazole;hexafluorophosphate
C1CCN(C1)C(=[N+]2CCCC2)ON3C4=CC=CC=C4N=N3.F[P-](F)(F)(F)(F)F
CAS Min % 97.5
CAS Max % 100.0
Melting Point 176°C
Color White
Elemental Analysis 40.73 to 42.81% C, 15.83 to 16.64% N
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 5 g
Formula Weight 431.32
Percent Purity 98%
Physical Form Powder
Chemical Name or Material O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate
Show More Show Less
compliance-icons
Product Identifier
  • O-(Benzotriazol-1-yl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'