missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Nylon Reducing Bushings

Sturdy threaded fittings are designed for permanent installation

Brand:  Masterflex™ 06349-31

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11741124

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Nylon Reducing Bushings
  • Available in a variety of matierials and sizes
  • Pack of Six
Specifications

Specifications

Reducing Bushing
1.03 in.
6 Fittings
Ethylene Oxide
0.79 in.
Nylon
26.19 mm
3/8 in. NPT(M) x 1/8 in. NPT(F)
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers