Learn More
Masterflex™ Nylon NPT(F) union elbows
Chemically-inert PFA handles most aggressive chemicals
Brand:  Masterflex™ 06349-64
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11781084
Quantity Price
1 60.59 EUR / Pack of 6
10 55.01 EUR / Pack of 6
Estimated Shipment
24-05-2021
Add to basket
Description
Description
  • Nylon NPT(F) union elbows
  • Available in a variety of matierials and sizes
  • Pack of Six
Specifications
Specifications
Nylon
60 mm
6 Fittings
Ethylene Oxide
Elbow-shaped
2.36 in.
PFA NPT(F) Union Elbows
3/4 in. NPT(F)
60 mm