missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Nylon NPT(F) union elbows

Chemically-inert PFA handles most aggressive chemicals

Brand:  Masterflex™ 06349-62

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11761084

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Nylon NPT(F) union elbows
  • Available in a variety of matierials and sizes
  • Pack of Six
Specifications

Specifications

PFA NPT(F) Union Elbows
1.42 in.
6 Fittings
Ethylene Oxide
1.42 in.
Nylon
36 mm
3/8 in. NPT(F)
36 mm
Elbow-shaped
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers