Learn More
Cole-Parmer™ Nylon Male Luer Fitting
Sterilizable male luer fitting available configured with a variety of fitting options. Cole-Parmer™ Nylon Male Luer Fitting has good chemical resistance.
Brand:  Cole-Parmer™ 45505-40
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15296333
  • 16.22 EUR / Pack of 25
Call for latest availability
+358 9 8027 6280
Add to basket
Description
Description
  • Good chemical resistance
  • Max temperature 455°F (235°C)
  • Sterilize by autoclave, ethylene oxide, or gamma irradiation
Specifications
Specifications
Nylon
Male Luer Fitting
Fits Male luer x 1/16 in. hose barb
25 Pack