Learn More
Sartorius™ Nutrient Media Pad Sets
Brand: Sartorius™ 14091--50------N
232.00 EUR valid until 2024-12-31
Use promo code "23270" to get your promotional price.
Additional Details : Weight : 0.01000kg
Specifications
Dehydrated Culture Media | |
Green | |
50 mm | |
50 mm |