Learn More
NUP210L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565465
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: PINSQLKLHTN REGAAFVSSR VLKCFPNSSV IEEDGEGLLK AGSIAGTAVL EVTSIEPFGV NQTTITGVQV APVTYLRVSS QPKLYTAQGR TLSAFPLGMS LTFTVQFYNS IGEKFHTHNT QLYLALN Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.
NUP210L Polyclonal antibody specifically detects NUP210L in Human samples. It is validated for Immunocytochemistry.Specifications
NUP210L | |
Polyclonal | |
Unconjugated | |
NUP210L | |
Nuclear pore membrane glycoprotein 210-like; nuclear pore membrane glycoprotein 210-like (LOC91181); Nucleoporin 210 kDa-like; nucleoporin 210 like; nucleoporin 210kDa like; nucleoporin 210kDa-like; nucleoporin 210-like; nucleoporin Nup210-like; NUP210L; RGD1310250 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
91181 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5VU65 | |
NUP210L | |
Recombinant Human NUP210L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only