Learn More
Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes
Open and close Thermo Scientific™ Nunc™ EZFlip™ Conical Centrifuge Tubes with one hand due to a proprietary hinged-cap system.
22.00 EUR - 328.00 EUR
Specifications
Material PP (Polypropylene)
Sterility Sterile
No. per Case 500
View More Specs
Products 6
Product Code Brand Capacity (Metric) Max. Force Packaging Price Quantity & Availability  
Product Code Brand Capacity (Metric) Max. Force Packaging Price Quantity & Availability  
10512802 View Documents Thermo Scientific™
362694
15mL 8,500 x g Bulk 216.00 EUR
Pack of 500 View alternative products
Save Up to 
»
In Stock 28
Add to basket
10571663 View Documents Thermo Scientific™
362697
50mL 9,500 x g Racked 328.00 EUR
Pack of 500 View alternative products
Save Up to 
»
Estimated Shipment
24-07-2019
Add to basket
10048631 View Documents Thermo Scientific™
362696
50mL 9,500 x g Bulk 241.00 EUR
Pack of 500 View alternative products
Save Up to 
»
In Stock 23
Add to basket
10777414 View Documents Thermo Scientific™
362695
15mL 8,500 x g Racked 255.00 EUR
Pack of 500 View alternative products
Save Up to 
»
Estimated Shipment
11-07-2019
Add to basket
11301645 View Documents Thermo Scientific™
362695
15mL 8,500 x g Racked 24.00 EUR
Pack of 50 View alternative products
Save Up to 
»
In Stock 7
Add to basket
12195700 View Documents Thermo Scientific™
362694
15mL 8,500 x g Bulk 22.00 EUR
Pack of 50 View alternative products
Save Up to 
»
Estimated Shipment
24-07-2019
Add to basket
Description
Description
Features:
  • Integrated leakproof cap prevents cross-contamination and lost closures
  • Conical bottom, sterile
  • Clarified polypropylene combines chemical resistance and high-speed tolerances
  • Use 50mL tubes up to 9500 x g; 15mL tubes withstand forces up to 8500 x g
  • White writing area and flat caps for sample identification
  • Printed volume markings and graduations
  • Standard dimensions
Ordering Information:
Tubes are sterile and are supplied in a bulk pack (sleeve) or a rack.
Warranty: 90 days
Specifications
Specifications
PP (Polypropylene)
Sterile
500
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.