Learn More
Thermo Scientific™ Nunc™ Deep Form Bacteriology Petri Dishes
Culture fungi, bacteria and other microorganisms with these petri dishes
12.10 EUR - 409.32 EUR
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
15300484
|
Thermo Scientific™
4014-12 |
150 mm |
249.00 EUR
Pack of 72 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
10765954
|
Thermo Scientific™
4031 |
100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15367036
|
Thermo Scientific™
4031 |
100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10581495
|
Thermo Scientific™
4036 |
60 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Culture fungi, bacteria and other microorganisms with Thermo Scientific™ Nunc Lab-Tek Disposable Petri Dishes.
Thermo Scientific Nunc Cell Culture products offer solutions for cell growth and analysis and have been used by researchers worldwide since 1953. With a wide range of surface areas and treatments, we have products for all your basic or advanced cell-based research.
Features:- Promote larger plaques used for efficient library screening
- Compatible with automated systems
- Sterile; disposable
- Deep Petri dishes allow for longer culture periods
Ordering Information:
12 per sealed plastic sleeve.
Specifications
Sterile |