missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NRSN1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550073
Additional Details : Weight : 0.01000kg
Description
The antibody detects endogenous levels of total NRSN1 protein.
NRSN1 Polyclonal antibody specifically detects NRSN1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
NRSN1 | |
Polyclonal | |
Unconjugated | |
NRSN1 | |
neurensin 1; neurensin-1; Neuro-p24; nrsn1; p24; vesicular membrain protein p24; vesicular membrane protein of 24 kDa; Vesicular membrane protein p24; Vmp | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140767, 22360, 291129 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P97799, Q8IZ57 | |
NRSN1 | |
Synthesized peptide derived from internal of human NRSN1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |