Learn More
Invitrogen™ Luminex™ MCP-1 Monkey Singleplex Bead Kit
Analyte specific components for the measurement of monkey MCP-1 (Monocyte Chemotactic/Chemoattracant Protein-1) in serum, plasma or tissue culture supernatant.
Brand: Invitrogen™ LPC1011
Additional Details : Weight : 0.05000kg
Includes: Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet.
Requires : Buffer reagents needed to complete the reaction sold separately (LMB0001).
Description
The assay may be run alone or in combination with other Monkey Singleplex Bead Kits from Invitrogen. Reagents intended for use in the Luminex 100™ or 200™ System only. EveryPrep™ Universal Vacuum Manifold available separately (Cat. No. K2111-01). Reagents intended for use in Luminex 100™ or 200™ and FLEXMAP 3D™ Systems only.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Room Temperature
Specifications
100 Assays | |
15 pg/mL | |
Luminex Instruments | |
NM_001032821.1, NP_001027993.1 | |
Cell Culture Supernatant, Plasma, Serum |
Polystyrene | |
Fluorescent Detection | |
574138 | |
MCP-1/CCL2 | |
Capture antibody coated beads, Detector antibody, Lot specific technical data sheet, Standard |
For Research Use Only. Not for use in diagnostic procedures.