Learn More
Invitrogen™ Luminex™ IFN-γ Human Singleplex Bead Kit
Analyte specific components for the measurement of human IFN-γ (Interferon-gamma) in serum, plasma or tissue culture supernatant
Brand: Invitrogen™ LHC4031
Additional Details : Weight : 0.06000kg
Includes: Contains capture antibody coated beads, standard, detector antibody and lot-specific technical data sheet.
Requires : Buffer reagents needed to complete the reaction sold separately (LHB0001).
Description
Assay may be run alone or in combination with other Human Singleplex Bead Kits. EveryPrep™ Universal Vacuum Manifold available separately (Cat. No. K2111-01). Reagents intended for use in Luminex 100™ or 200™ Systems only.
Cell Analysis, Cell Signaling, Cell Viability, Proliferation & Function, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet Ice
Specifications
100 Assays | |
<5 pg/mL | |
Luminex Instruments | |
NM_000619.2, NP_000610.2 | |
Cell Culture Supernatant, Plasma, Serum |
Polystyrene | |
Fluorescent Detection | |
3458 | |
IFN-γ | |
Capture antibody coated beads, Detector antibody, Lot specific technical data sheet, Standard |
For Research Use Only. Not for use in diagnostic procedures.