missing translation for 'onlineSavingsMsg'
Learn More

Merck™ Novagen™ rLysozyme™ Solution

For lysis of Gram-negative bacteria

Brand:  Merck™ 71110-3

Additional Details : Weight : 0.15000kg

 View more versions of this product

Product Code. 15285609

  • 66.72 EUR / 300ku


Call for latest availability
+358 9 8027 6280
Add to basket

Description

Description

  • Contains highly purified and stabilized recombinant lysozyme
  • Catalyzes hydrolysis of N-acetylmuramide linkages in bacterial cell walls
  • Specific activity of rLysozyme (1700KU/mg) for E. coli lysis is 250 times greater than that of chicken egg white lysozyme
  • Optimally active at physiological pH
Specifications

Specifications

Stabilized recombinant lysozyme
Supplied as a ready-to-use solution at a concentration range of 27 to 33KU/μL in 50% glycerol containing 50mM Tris-HCl, 0.1M NaCl, 0.1mM EDTA, 1 mM DTT, 0.1% TRITON™ X-100
300kU
27 to 33KU/μL
7.5
-20°C
Videos

For Research Use Only