Learn More
Merck™ Novagen™ BLR(DE3) Competent Cells
For production of protein from target genes cloned in pET vectors by induction with IPTG
Brand:  Merck™ 69053-4
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 15257644
  • 343.31 EUR / 1mL
Call for latest availability
+358 9 8027 6280
Add to basket
Description
Description
  • recA- derivative of BL21
  • Improves plasmid monomer yields
  • Helps stabilize target plasmids containing repetitive sequences or whose products may cause loss of DE3 prophage (1, 2)
Specifications
Specifications
1mL
BLR(DE3) Competent Cells
≤ -70°C