missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOTCH3, APC, clone: HMN3-133, eBioscience™
Armenian Hamster Monoclonal Antibody
Brand: Affymetrix eBioscience 17-5763-80
Additional Details : Weight : 0.25000kg
Description
Description: This HMN3-133 monoclonal antibody reacts with mouse Notch3, one of four members of the Notch family of receptors. Notch receptors are 300-kDa single-pass transmembrane proteins. While the extracellular domain contains numerous epidermal growth factor-like repeats for ligand binding, the intracellular domain is involved in cell signaling. Upon binding its membrane-bound ligand (either Delta or Jagged), the Notch receptor undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, heterodimerizes with the DNA-binding protein RBP-J, recruits co-activator molecules, and ultimately activates transcription.Notch 3 expression has been demonstrated on regulatory T cells and some thymocyte subsets, including CD4-CD8- and CD8SP cells. In addition, this Notch receptor is prevalent in vascular smooth muscle and the central nervous system. In addition to its role in stem cell hematopoiesis, Notch 3 plays a pivotal role in mammalian T cell lineage commitment and thymocyte development. Specifically, Notch 3 has been implicated in the CD4-CD8- double negative (DN) to CD4+CD8+ double positive transition with greatest expression in DN3 (CD4-CD8-CD25+CD44-) cells. Notch 3 may also be involved in regulating the pre-TCR checkpoint via crosstalk with the NFkappaB and E2A signaling pathways. Finally, overexpression of Notch 3 has been implicated in leukemia.Applications Reported: This HMN3-133 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HMN3-133 antibody has been tested by flow cytometric analysis of mouse thymocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.Specifications
Notch3 | |
Monoclonal | |
0.2mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
Q61982 | |
CADASIL, CASIL | |
Armenian Hamster | |
Affinity chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Murine |
Flow Cytometry | |
HMN3-133 | |
APC | |
Notch3 | |
Conjugated | |
Notch3 | |
IgG | |
25μg | |
Store at 2-8°C. Do not freeze. Light-sensitive material. | |
Primary | |
18131 |
For Research Use Only.