Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Increase pipetting performance with Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency.
Brand:  Thermo Scientific™ 918-262-05
UNSPSC : 41121608
Additional Details : Weight : 1.62000kg
 View more versions of this product
Product Code. 10008321
  • 95.30 EUR / Pack of 960
Call for latest availability
+46 (0)31 352 32 30
Add to basket
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
  • Available in low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384- and 1536- deep well plates
  • Low percentage coefficient of variation (%CV)
Description & Specifications
Specifications
20μL
Nonsterile
96 Tips/Rack
Beckman Biomek FX/NX and 3000 and Multimek AP96 Workstations
No
Clear
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Beckman