Learn More
Brand™ Non-Sterile Polyethylene Filter Tips
Non-self-sealing filter tips from Brand™ have a PE filter that is free from chemical additives.
Brand:  Brand™ 732612
84.30 EUR valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Additional Details : Weight : 1.04800kg
 View more versions of this product
Product Code. 15154957
  • 92.00 EUR / Pack of 960
Estimated Shipment: 05-06-2024
Description
  • Permeability is controlled by the combination of pore size and filter length, so that no aerosols can reach the pipet shaft
  • Functions with consistent reliability
  • Sample can be recovered from the filter by simply actuating the pipette’s blow-out function, or by centrifugation if necessary
  • Available in a wide range of capacities
Specifications
Specifications
5 to 200 μL
Non-sterile
Ideal for biological samples that contain detergents such as TRITON™ X-100, SDS, Tween
Filter Tip
960
Polyethylene
Filtered
Yellow
Rack
Racked: 10 TipRacks, 96 each