missing translation for 'onlineSavingsMsg'
Learn More

Nitric Acid solution 0.1mol/L, Fisher Chemical™

Catalog Number 12920764
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 0.1M (0.1N)
CAS 7697-37-2
Packaging Bottle
Quantity 100 mL
Molecular Formula HNO3
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Concentration or Composition (by Analyte or Components) 0.1mol/L
Chemical Name or Material Nitric Acid solution
Show More Show Less
compliance-icons
Product Identifier
  • Nitric acid 1M Volumetric Solution (SOL 118)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'