Learn More
R&D Systems NHP XL CXCL10/IP-10, Magnetic Luminex™ Performance Assay
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Quantity | 1 Kit |
| Species | Primate |
| Binding Target | CXCL10/IP-10/CRG-2 |
| Gene ID (Entrez) | 3627 |
| Synonym | C7, chemokine (C-X-C motif) ligand 10, crg-2, CRG2, gIP-10, IFI10, INP10, IP-10, mob-1, SCYB10 |
| Content And Storage | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |