Learn More
Lovibond™ Nessleriser 2150 Water Testing System
Brand:  Lovibond™ 172150
Additional Details : Weight : 1.70000kg
Product Code. 13111671
  • 290.00 EUR / Each
Estimated Shipment: 14-06-2024
Specifications
Uses natural light and cylinders with a path length of 113 mm and a cell capacity of 50mL