Learn More
Fisherbrand™ Neoprene Tubing
Used in flow lines for fluids and gases in a variety of applications.
16.60 EUR - 473.00 EUR
Specifications
Material Neoprene
Overall Length (Metric) 10 m
View More Specs
Products 13
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
11885492
View Documents Promotion Details
Fisherbrand™
10 x 12mm
52.15 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
11577063
View Documents Promotion Details
Fisherbrand™
10 x 24mm
217.00 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
11895492
View Documents Promotion Details
Fisherbrand™
12.5 x 17mm
65.70 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
11587063
View Documents Promotion Details
Fisherbrand™
12.5 x 29.5mm
473.00 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
11825502
View Documents Promotion Details
Fisherbrand™
25 x 31.5mm
183.00 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
11845492
View Documents Promotion Details
Fisherbrand™
3 x 4.5mm
16.60 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
11835502
View Documents Promotion Details
Fisherbrand™
3 x 9mm
39.40 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 28-06-2024
to see stock.
Add to basket
11845502
View Documents Promotion Details
Fisherbrand™
5 x 13mm
74.75 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
11855492
View Documents Promotion Details
Fisherbrand™
5 x 8mm
24.45 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
10366344
View Documents Promotion Details
Fisherbrand™
6.5 x 1.5mm
26.05 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
11875492
View Documents Promotion Details
Fisherbrand™
8 x 10mm
41.00 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Add to basket
11865502
View Documents Promotion Details
Fisherbrand™
8 x 20mm
160.00 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
11855502
View Documents Promotion Details
Fisherbrand™
6.5 x 10mm
119.00 EUR
10m
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
Description
  • Resistant to heat, ozone, petroleum oils and hydrocarbon solvents
  • Temperature range: -30°C to 100°C
Specifications
Specifications
Neoprene
10 m