Learn More
Fisherbrand™ Solid Neoprene Stoppers
Offer resistance to heat, ozone and petroleum based oils.
4.80 EUR - 23.25 EUR
Specifications
Color Gray
Material Neoprene
Product Type Solid Stopper
View More Specs
Products 11
Product Code Brand Diameter (Metric) Bottom Height (Metric) Stopper Number Diameter (Metric) Top Price Quantity & Availability  
Product Code Brand Diameter (Metric) Bottom Height (Metric) Stopper Number Diameter (Metric) Top Price Quantity & Availability  
11587103
View Documents Promotion Details
Fisherbrand™
37 mm 38.5mm 37 42.5 mm
19.20 EUR
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
11577093
View Documents Promotion Details
Fisherbrand™
15 mm 24mm 15 18 mm
10.30 EUR
Pack of 20
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
11557103
View Documents Promotion Details
Fisherbrand™
31 mm 35mm 31 36 mm
13.85 EUR
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
11577103
View Documents Promotion Details
Fisherbrand™
35 mm 38.5mm 35 40.5 mm
17.75 EUR
Pack of 5
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
11507113
View Documents Promotion Details
Fisherbrand™
41 mm 42mm 41 47 mm
12.55 EUR
Pack of 2
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
11597083
View Documents Promotion Details
Fisherbrand™
5 mm 16mm 5 7 mm
4.80 EUR
Pack of 20
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
11527113
View Documents Promotion Details
Fisherbrand™
45 mm 42mm 45 51 mm
13.00 EUR
Pack of 2
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
11527093
View Documents Promotion Details
Fisherbrand™
8 mm 20mm 8 10.5 mm
6.15 EUR
Pack of 20
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
11507103
View Documents Promotion Details
Fisherbrand™
21 mm 28mm 21 24.5 mm
9.70 EUR
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
to see stock.
Add to basket
11517103
View Documents Promotion Details
Fisherbrand™
23 mm 28mm 23 26.5 mm
11.90 EUR
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 13-06-2024
to see stock.
Add to basket
11547103
View Documents Promotion Details
Fisherbrand™
29 mm 32mm 29 33 mm
23.25 EUR
Pack of 10
View alternative products
Save Up to 
»
Estimated Shipment: 27-06-2024
Description
  • Better resistance to heat, ozone and petroleum based oils than red rubber
  • Less suitable for low temperature use
  • Operational temperature range -30°C to 100°C
Specifications
Specifications
Gray
Neoprene
Solid Stopper