Learn More
Trajan Scientific and Medical™ Needle 5-10MA(N)WLC replacement HPLC tip 5mL to 10mL 2 x 0.028 inch needle length x outer diameter SGE
A range of quality needles to fit a SGE syringe in a range of lengths and tip styles
Brand:  Trajan Scientific and Medical™ 0315234
Additional Details : Weight : 0.50000kg
Product Code. 12927616
  • 49.50 EUR / Pack of 5
Estimated Shipment: 15-07-2024
Description
  • Needle length is exposed length of needle
  • Choose different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
24 G
0.2 mm
0.5 mm
LC