missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Ndelta-Benzyloxycarbonyl-L-ornithine, 98%

Brand:  Alfa Aesar™ H62435.03

Additional Details : CAS Number : 3304-51-6 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458938

  • 53.80 EUR / 1g


Estimated Shipment date
from Supplier 24-04-2019
Add to basket

Specifications

Chemical Identifiers

3304-51-6
266.297
VULSXQYFUHKBAN-NSHDSACASA-N
7009570
C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)N
C13H18N2O4
MFCD00038200
h-orn z-oh, ndelta-cbz-l-ornithine, n'-cbz-l-ornithine, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, n-delta-benzyloxycarbonyl-l-ornithine, ornithine, n5-phenylmethoxy carbonyl, 2s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, h-orn cbz-oh, ambotzhaa1227, n, a-cbz-l-ornithine
(2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoic acid

Specifications

Ndelta-Benzyloxycarbonyl-L-ornithine
C13H18N2O4
h-orn z-oh, ndelta-cbz-l-ornithine, n'-cbz-l-ornithine, s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, n-delta-benzyloxycarbonyl-l-ornithine, ornithine, n5-phenylmethoxy carbonyl, 2s-2-amino-5-benzyloxy carbonyl amino pentanoic acid, h-orn cbz-oh, ambotzhaa1227, n, a-cbz-l-ornithine
C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)N
266.297
266.29
98%
3304-51-6
MFCD00038200
VULSXQYFUHKBAN-NSHDSACASA-N
(2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoic acid
7009570
Solid
1g
Videos
SDS