Learn More
NAV3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556932
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PSQSLSKPIT MEKASASSCP APLEGREAGQ ASPSGSCTMT VAQSSGQSTG NGAVQLPQQQ QHSHPNTATV APFIYRAHSE NEGTALPSAD S Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
NAV3 Polyclonal antibody specifically detects NAV3 in Human samples. It is validated for Immunohistochemistry.Specifications
NAV3 | |
Polyclonal | |
Unconjugated | |
NAV3 | |
4732483H20Rik; 9630020C08Rik; KIAA0938; mKIAA0938; NAV3; Neuron navigator 3; POMFIL1; Pomfil1p; pore membrane and/or filament interacting like protein 1; pore membrane and/or filament-interacting-like protein 1; RGD1306259; steerin 3; STEERIN3; Steerin-3; Unc-53 homolog 3; unc53H3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89795 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IVL0 | |
NAV3 | |
Recombinant protein corresponding to Human NAV3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only