Learn More
Ark Plas Products™ Natural Polypropylene Commercial Grade Barbed Straight Connectors
Commercial Grade Fittings are designed as a low-cost alternative with high-end performance for tubing connections in the laboratory.
Brand: Ark Plas Products™ CAC59-PR0
Additional Details : Weight : 0.06300kg
Description
- Economic with reliable performance
- Pressure rating up to 125 P.S.I. without clamps
- Contain recycled runner material
Specifications
Natural | |
Polypropylene | |
25 Pack | |
Straight |
Autoclavable | |
Barbed Straight Connector | |
3/8 x 3/8 in. Barb |