missing translation for 'onlineSavingsMsg'
Learn More

Ark Plas Products™ Natural Polypropylene Barbed Elbow Connectors

A fluted polypropylene connection for flexible tubes in the laboratory.

Brand:  Ark Plas Products™ CDA57-PR0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11751956

  • 134.18 EUR / Pack of 25


Estimated Shipment
14-04-2021
Add to basket

Description

Description

  • Offers excellent chemical resistance
  • Commercial grade
  • Operating temperature from −20° to +135°C
Specifications

Specifications

Natural
Polypropylene
25 Pack
Autoclavable
Barbed Elbow Connector
3/16 x 3/16 in. Barb
Videos
SDS
Documents