missing translation for 'onlineSavingsMsg'
Learn More

500GR Sodium Iodide BAKER

Brand:  JT Baker 0294.0500

309.70 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.



500GR Sodium Iodide BAKER

Additional Details : CAS Number : 7681-82-5 Weight : 0.50000kg


Product Code. 10294400

  • 326.00 EUR / 500g
Estimated Shipment: 15-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 500GR Sodium Iodide BAKER

Signal Word
  • Danger

Hazard Category
  • Specific target organ toxicity Category 1

Hazard Statement
  • H372-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium iodide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers