Learn More
Thermo Scientific™ National Target DP™ Screw Thread Caps
Thermo Scientific™ National Target DP Screw Thread Caps with open top are designed for use with Target DP septa.
Brand:  Thermo Scientific™ C5000-98Y
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 15286543
  • 5.76 EUR / Pack of 100
Estimated Shipment
07-05-2019
Add to basket
Description
Description
  • Polypropylene, open-top caps for use with Target DP septa
Specifications
Specifications
Yellow
Yellow
National Target DP™
Screw Cap
Polypropylene
Target DP septa
No Septum
Videos