Learn More
Buerkle™ Narrow Neck Reagent Bottle
2.37 EUR valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Use promo code "23267" to get your promotional price.
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
Height (Metric):
108 mm
136 mm
170 mm
Beschreibung
- HDPE, transparent
- PP tamper-evident screw cap, blue
Spezifikation
Product Type | Reagent Bottle |
Material | HDPE |
Includes | Polypropylene tamper-evident blue screw cap |
Closure Material | Polypropylene |
Capacity (Metric) | 100 mL |
Color | Transparent |
For Use With (Application) | Suitable for Viscous Oils |
Height (Metric) | 108 mm |
Length (Metric) | 45 mm |
Width (Metric) | 45 mm |