missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Reagent Bottle, Narrow Neck, Clear, Without Stopper
252.70 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "23258" to get your promotional price.
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
5000 mL
Description
The classical stoppered reagent bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the storage of liquid and solid chemicals and reagents. The bottles feature a narrow conical ground joint neck and comply with ISO 4796-2. The narrow neck is suitable for liquids and the angle of the shoulders ensures that the contents can be smoothly poured. The glass stopper provides an excellent seal that protects the contents from evaporation, oxidation, and contamination. The bottles have a Retrace Code for manufacturing lot traceability and a marking field in permanent white enamel.
- All borosilicate glass components and therefore suitable for storage of aggressive materials
- Tight closure due to standard ground joint
- Easy-to-read scale with graduation marks to estimate the content
- Fired-on, highly durable white ceramic print
Specifications
Diameter (Metric) | 42 mm |
Capacity (Metric) | 50 mL |
Description | Neck with standard ground joint |
For Use With (Application) | Storage of air sensitive reagents, pharmaceuticals, and chemicals; Storage of hygroscopic liquids and pharmaceuticals; Transport and shipping of samples; Storage of hygroscopic liquids and pharmaceuticals |
Material | Borosilicate Glass (3.3) |
Quantity | 10/Pk. |
Autoclavable | Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'