missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-O-tert-butyl-L-threonine, 98%, Thermo Scientific Chemicals

Catalog Number p-7040142
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
71989-35-0
C23H27NO5
397.47
MFCD00077075
LZOLWEQBVPVDPR-VLIAUNLRSA-N
fmoc-thr tbu-oh, fmoc-o-tert-butyl-l-threonine, fmoc-thr t-bu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, fmoc-thr but, 2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine, fmoc-thr but-oh, fmoc-o-tert.butyl-l-threonine, fmoc-thr otbu-oh
6364643
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
This item is not returnable. View return policy
71989-35-0
C23H27NO5
397.47
MFCD00077075
LZOLWEQBVPVDPR-VLIAUNLRSA-N
fmoc-thr tbu-oh, fmoc-o-tert-butyl-l-threonine, fmoc-thr t-bu-oh, n-9-fluorenylmethoxycarbonyl-o-tert-butyl-l-threonine, fmoc-thr but, 2s,3r-3-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino butanoic acid, n-9h-fluoren-9-yl methoxy carbonyl-o-tert-butyl-l-threonine, fmoc-thr but-oh, fmoc-o-tert.butyl-l-threonine, fmoc-thr otbu-oh
6364643
(2S,3R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]butanoic acid
C[C@@H](OC(C)(C)C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 131°C to 134°C
Quantity 2 g
Beilstein 4581133
Optical Rotation +16° (c=1 in Ethyl Acetate)
Formula Weight 397.46
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-O-tert-butyl-L-threonine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'