missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-D-arginine, 98%, Thermo Scientific™

Catalog Number 15485575
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
187618-60-6
C34H40N4O7S
648.78
MFCD00237010
HNICLNKVURBTKV-UHFFFAOYNA-N
fmoc-d-arg pbf-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, 2r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-yl sulfonyl carbamimidamido pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, ambotzfaa1305, fmoc-darg pbf-oh, pubchem12397, na-fmoc-n?-pbf-d-arginine, fmoc-n-guanidino-pbf-d-arginine, nalpha-fmoc-nomega-pbf-d-arginine
46737258
CC1=C2CC(C)(C)OC2=C(C)C(C)=C1S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
This item is not returnable. View return policy
187618-60-6
C34H40N4O7S
648.78
MFCD00237010
HNICLNKVURBTKV-UHFFFAOYNA-N
fmoc-d-arg pbf-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, 2r-2-9h-fluoren-9-yl methoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-2,3-dihydro-1-benzofuran-5-yl sulfonyl carbamimidamido pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, ambotzfaa1305, fmoc-darg pbf-oh, pubchem12397, na-fmoc-n?-pbf-d-arginine, fmoc-n-guanidino-pbf-d-arginine, nalpha-fmoc-nomega-pbf-d-arginine
46737258
CC1=C2CC(C)(C)OC2=C(C)C(C)=C1S(=O)(=O)NC(N)=NCCCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 9537282
IUPAC Name (2R)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
Formula Weight 648.77
Percent Purity 98%
Chemical Name or Material Nalpha-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-D-arginine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'