missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Nalpha-Boc-Nomega-nitro-L-homoarginine, 98%

Brand:  Alfa Aesar™ H62760.06

Additional Details : CAS Number : 28968-64-1 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15459238

  • 807.00 EUR / 5g


Estimated Shipment date
from Supplier 24-04-2019
Add to basket

Specifications

Chemical Identifiers

28968-64-1
333.345
YFIPSHDRWHZYOM-QMMMGPOBSA-N
26771621
C12H23N5O6
MFCD00151901
boc-homoarg no2-oh, boc-hoarg no2-oh, boc-harg no2, boc-n'-nitro-l-homoarginine, s-2-tert-butoxycarbonyl amino-6-3-nitroguanidino hexanoic acid, 2s-2-tert-butoxycarbonyl amino-6-n'-nitrocarbamimidamido hexanoic acid, boc-homoarg no-oh, boc-hag no2-oh, pubchem18972, boc-harg no2-oh
CC(C)(C)OC(=O)NC(CCCCN=C(N)N[N+](=O)[O-])C(=O)O

Specifications

Nalpha-Boc-Nomega-nitro-L-homoarginine
C12H23N5O6
boc-homoarg no2-oh, boc-hoarg no2-oh, boc-harg no2, boc-n'-nitro-l-homoarginine, s-2-tert-butoxycarbonyl amino-6-3-nitroguanidino hexanoic acid, 2s-2-tert-butoxycarbonyl amino-6-n'-nitrocarbamimidamido hexanoic acid, boc-homoarg no-oh, boc-hag no2-oh, pubchem18972, boc-harg no2-oh
CC(C)(C)OC(=O)NC(CCCCN=C(N)N[N+](=O)[O-])C(=O)O
26771621
Solid
5g
28968-64-1
MFCD00151901
YFIPSHDRWHZYOM-QMMMGPOBSA-N
333.345
333.34
98%
Videos
SDS