missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.
Solubility
Soluble in dilute acid.
Notes
Hygroscopic. Store in cool, well ventilated area. Keep container tightly closed. Store away from water/moisture.
Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid is a important building block for the synthesis of peptides containing DAP residues, e.g. bleomycins, edeines, tuberactinomycins.
Solubility
Soluble in dilute acid.
Notes
Hygroscopic. Store in cool, well ventilated area. Keep container tightly closed. Store away from water/moisture.
Specifications
Melting Point | ∼240°C (decomposition) |
Beilstein | 4486950 |
Solubility Information | Soluble in dilute acid. |
Formula Weight | 238.24 |
Percent Purity | 98% |
Quantity | 1 g |
Optical Rotation | −39° (c-1 in 1M HCl) |
Sensitivity | Hygroscopic |
Chemical Name or Material | Nalpha-Benzyloxycarbonyl-L-2,3-diaminopropionic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'