Learn More
Thermo Scientific™ Nalgene™ HDPE Quick Disconnects
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ Quick Disconnects.
Brand: Thermo Scientific™ 6150-0020
Additional Details : Weight : 0.07020kg
Description
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ HDPE Quick Disconnects, which allow tubulations to slide together snugly with a twist.
Two tapered tubulations fit tightly together with male/female connection, allowing for rapid joining or separation of apparatus lines.Specifications
Quick Disconnect | |
HDPE | |
3/8 to 7/16 in. I.D. | |
12/Pk., 72/Cs. |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.