missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Autoclavable Polypropylene Carboys with Bottom Tubulation

Ensure more complete drainage with the low outlet of these polypropylene carboys, excellent for distilled water and culture media.

52.00 EUR - 88.50 EUR

Specifications

Description Autoclavable Carboy w/Bottom Tubulation
Diameter (Metric) Inner Neck 64 mm
Autoclavable Yes
Closure Size 83 B
Graduations Molded-in graduations in 1 gal. and 5 L increments
View More Specs

Alert:

Completely disengage threads or remove closure before autoclaving. Not recommended for hazardous materials.


Products 2
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Size Price Quantity & Availability  
10461103 View accessory products View Documents Thermo Scientific™
2301-0020
10 L 83 B 52.00 EUR
Each View alternative products
Save Up to 
»
Estimated Shipment
04-12-2019
Add to basket
 
10550363 View accessory products View Documents Thermo Scientific™
2301-0050
20 L 83 B 88.50 EUR
Each View alternative products
Save Up to 
»
Estimated Shipment
04-12-2019
Add to basket
 
Description

Description

Features:
  • Bottom tubulation molded in one piece with bottle
  • Molded-in handles for ease in transporting containers
  • Molded-in graduations in 1 gal. and 5L increments
  • Neck accepts No. 13.5 rubber stoppert
  • Autoclavable/Graduated/Leakproof†

Ordering Information:

Both sizes accept 0.5in. I.D. (1.3cm) tubing - Not Included

Includes:

White polypropylene screw cap with TPE gasket

Note:

A hose clamp should be used when attaching tubing to ensure tight fit. Completely disengage threads or remove closure before autoclaving. Not recommended for hazardous materials.

Warranty: 90 days.  For warranty terms, please contact technicalsupport@thermofisher.com

Specifications

Specifications

Autoclavable Carboy w/Bottom Tubulation
Yes
Molded-in graduations in 1 gal. and 5 L increments
Polypropylene
64 mm
83 B
Molded-in handles
1

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.