Learn More
NAALADL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561434
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: RLSDSFFLPL KVSDYSETLR SFLQAAQQDL GALLEQHSIS LGPLVTAVEK FEAEAAALGQ RISTLQKGSP DPLQVRMLN Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 82%.
NAALADL1 has NAALADase-like activity which is currenty unknown in function. NAALADL1 has no NAAG hydrolyzing activity, exhibits a dipeptidyl-peptidase IV type activity. In vitro, cleaves Gly-Pro-AMC.Specifications
NAALADL1 | |
Polyclonal | |
Unconjugated | |
Naaladl1 | |
100 kDa ileum brush border membrane protein; Aminopeptidase NAALADL1; Gm964; I100; ileal dipeptidylpeptidase; ileal peptidase I100; NAALADase L; NAALADASEL; NAALADase-like 1; NAALADL; Naaladl1; N-acetylated alpha-linked acidic dipeptidase-like 1; N-acetylated-alpha-linked acidic dipeptidase like protein; N-acetylated-alpha-linked acidic dipeptidase-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10004 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UQQ1 | |
Naaladl1 | |
Recombinant protein corresponding to Human NAALADL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only