missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylglycine hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number p-7055639
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
6939-23-7
C6H14ClNO2
167.63
MFCD03788441
QORBVLNEGUIPND-UHFFFAOYSA-N
2-tert-butylamino acetic acid hydrochloride, n-tert-butylglycine hydrochloride, n-t-butylglycine hydrochloride, n-t-butylglycine hcl, unii-b8y856vh2e, t-butylglycine hydrochloride, glycine, n-1,1-dimethylethyl-, hydrochloride 1:1, glycine, n-1,1-dimethylethyl-, hydrochloride, t-butylaminoacetic acid hydrochloride, glycine, n-tert-butyl-, hydrochloride
17748195
hydrogen 2-(tert-butylamino)acetic acid chloride
[H+].[Cl-].CC(C)(C)NCC(O)=O
This item is not returnable. View return policy
6939-23-7
C6H14ClNO2
167.63
MFCD03788441
QORBVLNEGUIPND-UHFFFAOYSA-N
2-tert-butylamino acetic acid hydrochloride, n-tert-butylglycine hydrochloride, n-t-butylglycine hydrochloride, n-t-butylglycine hcl, unii-b8y856vh2e, t-butylglycine hydrochloride, glycine, n-1,1-dimethylethyl-, hydrochloride 1:1, glycine, n-1,1-dimethylethyl-, hydrochloride, t-butylaminoacetic acid hydrochloride, glycine, n-tert-butyl-, hydrochloride
17748195
hydrogen 2-(tert-butylamino)acetic acid chloride
[H+].[Cl-].CC(C)(C)NCC(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 223°C to 224°C
Color White
Linear Formula (CH3)3CNHCH2COOH·HCl
Beilstein 6296616
Formula Weight 167.63
Percent Purity 97%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-tert-Butylglycine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'