missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Phenylmaleimide, 98+%, Thermo Scientific Chemicals
39.86 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
500 g
Description
N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.
Solubility
Soluble in water (slightly ), methanol, ethanol, and benzene.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
N-Phenylmaleimide is used as a dipolarophile in 1,3-cycloaddition reaction with a nitrone, useful for the preparation of crystalline adducts of dienes. It is used as a plastic modifier.
Solubility
Soluble in water (slightly ), methanol, ethanol, and benzene.
Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Specifications
Melting Point | 87°C to 91°C |
Boiling Point | 162°C to 163°C (12 mmHg) |
Quantity | 25 g |
UN Number | UN2811 |
Beilstein | 125098 |
Merck Index | 14,7299 |
Solubility Information | Soluble in water (slightly ),methanol,ethanol,and benzene. |
Formula Weight | 173.17 |
Percent Purity | ≥98% |
Chemical Name or Material | N-Phenylmaleimide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'