Learn More
N,N-Dimethylglycine hydrochloride, 99%, Thermo Scientific Chemicals
Description
N,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. Used in the development of glycine-based ionic liquids and emulsifiers. Also it is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N-Dimethylglycine (DMG) is a natural N-methylated glycine that is used in comparative analysis with other N-methylated glycines such as sarcosine and βine. Used in the development of glycine-based ionic liquids and emulsifiers. Also it is potentially useful as a biomarker of protein degradation in COPD patients. DMG is used as a substrate to identify, differentiate and characterize amino acid methyltransferase(s).
Solubility
Soluble in water (almost transparency).
Notes
Hygroscopic, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Melting Point | 189°C to 194°C |
Molecular Formula | C7H4BrNO |
Quantity | 25 g |
Beilstein | 3910578 |
Sensitivity | Hygroscopic |
Merck Index | 14,3244 |
Synonym | 4-bromophenyl isocyanate, 4-bromophenylisocyanate, p-bromophenyl isocyanate, benzene, 1-bromo-4-isocyanato, p-bromophenylisocyanate, p-bromocarbanil, p-bromophenyl carbonimide, isocyanic acid 4-bromophenyl ester, unii-mgr77m31zw, mgr77m31zw |
Solubility Information | Soluble in water (almost transparency). |
InChI Key | CZQIJQFTRGDODI-UHFFFAOYSA-N |
SMILES | C1=CC(=CC=C1N=C=O)Br |
Show More |
RUO – Research Use Only