missing translation for 'onlineSavingsMsg'
Learn More

N,N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, free acid, MP Biomedicals™

Catalog Number p-4403862
Click to view available options
Quantity:
100 g
250 g
1 kg
This item is not returnable. View return policy
10191-18-1
C6H15NO5S
213.25
MFCD00007533
AJTVSSFTXWNIRG-UHFFFAOYSA-N
N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, N, N-Bis(2-hydroxyethyl)taurine
73243
CHEBI:39045
2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
OCCN(CCO)CCS(O)(=O)=O
This item is not returnable. View return policy
10191-18-1
C6H15NO5S
213.25
MFCD00007533
AJTVSSFTXWNIRG-UHFFFAOYSA-N
N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, N, N-Bis(2-hydroxyethyl)taurine
73243
CHEBI:39045
2-[bis(2-hydroxyethyl)amino]ethane-1-sulfonic acid
OCCN(CCO)CCS(O)(=O)=O
Color White
Physical Form Crystalline Powder
Quantity 100 g
Assay Percent Range 0.9961
Formula Weight 239.23
Chemical Name or Material N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, free acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'