missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-(3-methyl-1,2,4-oxadiazol-5-yl)benzylamine, 95%, Thermo Scientific™

Catalog Number p-3586875
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
921938-59-2
C11H13N3O
203.245
MFCD09817473
FWMVARCHFJQGJP-UHFFFAOYSA-N
n-methyl-3-3-methyl-1,2,4-oxadiazol-5-yl benzylamine, methyl 3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, 3-methyl-5-3-methylamino methyl phenyl-1,2,4-oxadiazole, methyl 3-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
24229501
N-methyl-1-[3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
CC1=NOC(=N1)C2=CC(=CC=C2)CNC
This item is not returnable. View return policy
921938-59-2
C11H13N3O
203.245
MFCD09817473
FWMVARCHFJQGJP-UHFFFAOYSA-N
n-methyl-3-3-methyl-1,2,4-oxadiazol-5-yl benzylamine, methyl 3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-3-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, 3-methyl-5-3-methylamino methyl phenyl-1,2,4-oxadiazole, methyl 3-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
24229501
N-methyl-1-[3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
CC1=NOC(=N1)C2=CC(=CC=C2)CNC
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material N-methyl-3-(3-methyl-1, 2, 4-oxadiazol-5-yl)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'