missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-(1,3-thiazol-2-yl)benzylamine, 95%, Thermo Scientific™

Catalog Number p-3522603
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
892501-89-2
C11H12N2S
204.291
MFCD09025830
MNTPWZTYKCKBQX-UHFFFAOYSA-N
n-methyl-3-1,3-thiazol-2-yl benzylamine, n-methyl-1-3-thiazol-2-yl phenyl methanamine, methyl 3-1,3-thiazol-2-yl phenyl methyl amine, benzenemethanamine,n-methyl-3-2-thiazolyl, n-methyl-1-3-1,3-thiazol-2-yl phenyl methanamine
18525729
N-methyl-1-[3-(1,3-thiazol-2-yl)phenyl]methanamine
CNCC1=CC=CC(=C1)C2=NC=CS2
This item is not returnable. View return policy
892501-89-2
C11H12N2S
204.291
MFCD09025830
MNTPWZTYKCKBQX-UHFFFAOYSA-N
n-methyl-3-1,3-thiazol-2-yl benzylamine, n-methyl-1-3-thiazol-2-yl phenyl methanamine, methyl 3-1,3-thiazol-2-yl phenyl methyl amine, benzenemethanamine,n-methyl-3-2-thiazolyl, n-methyl-1-3-1,3-thiazol-2-yl phenyl methanamine
18525729
N-methyl-1-[3-(1,3-thiazol-2-yl)phenyl]methanamine
CNCC1=CC=CC(=C1)C2=NC=CS2
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 204.3
Percent Purity 95%
Chemical Name or Material N-Methyl-3-(1, 3-thiazol-2-yl)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'