missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-benzyl-L-tyrosine, 98%, Thermo Scientific™

Catalog Number p-7039904
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
71989-40-7
C31H27NO5
493.56
MFCD00065682
REHSJSKPWIOKIJ-KSTXGFNDNA-N
fmoc-tyr bzl-oh, fmoc-o-benzyl-l-tyrosine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, l-tyrosine, n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl, fmoc-tyr bzl, 2s-3-4-benzyloxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-o-benzyltyrosine, ambotzfaa1754, pubchem19060, n-fmoc-o-benzyl-l-tyrosine
14427834
3-[4-(benzyloxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
OC(=O)[C@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2
This item is not returnable. View return policy
71989-40-7
C31H27NO5
493.56
MFCD00065682
REHSJSKPWIOKIJ-KSTXGFNDNA-N
fmoc-tyr bzl-oh, fmoc-o-benzyl-l-tyrosine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzyloxy phenyl propanoic acid, l-tyrosine, n-9h-fluoren-9-ylmethoxy carbonyl-o-phenylmethyl, fmoc-tyr bzl, 2s-3-4-benzyloxy phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-o-benzyltyrosine, ambotzfaa1754, pubchem19060, n-fmoc-o-benzyl-l-tyrosine
14427834
3-[4-(benzyloxy)phenyl]-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
OC(=O)[C@H](CC1=CC=C(OCC2=CC=CC=C2)C=C1)NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 157°C to 161°C
Quantity 1 g
Beilstein 4341195
Optical Rotation −16° (c=1 in DMF)
Formula Weight 493.55
Percent Purity 98%
Chemical Name or Material N-Fmoc-O-benzyl-L-tyrosine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'