missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Ethyl-N-phenylbenzenesulfonamide, 97%

Brand:  Alfa Aesar™ H60851.03

Additional Details : CAS Number : 54437-69-3 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15487348

  • 646.00 EUR / 1g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

N-Ethyl-N-phenylbenzenesulfonamide
261.34
3141147
54437-69-3
MFCD00590965

Specifications

N-Ethyl-N-phenylbenzenesulfonamide
Liquid
261.34
3141147
1g
54437-69-3
97%
MFCD00590965
Yellow
Videos
SDS