missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-S-benzyl-D-cysteine, 97%

Brand:  Alfa Aesar™ H62724.03

Additional Details : CAS Number : 102830-49-9 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15459208

  • 61.60 EUR / 1g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

102830-49-9
311.396
IFVORPLRHYROAA-GFCCVEGCSA-N
78754
CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(=O)O
C15H21NO4S
MFCD00038253
n-boc-s-benzyl-d-cysteine, boc-d-cys bzl-oh, boc-s-benzyl-d-cysteine, n-tert-butyloxycarbonyl-s-benzylcysteine, 2s-3-benzylsulfanyl-2-tert-butoxycarbonyl amino propanoic acid, boc-s-benzyl-d-cys, boc-s-benzyl-l-cys, ambotzbaa5410, s-benzyl-n-tert-butoxycarbonyl-d-cysteine, alanine, 3-benzylthio-n-carboxy-, n-tert-butyl ester, l
(2S)-3-benzylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

N-Boc-S-benzyl-D-cysteine
C15H21NO4S
5758339
IFVORPLRHYROAA-GFCCVEGCSA-N
(2S)-3-benzylsulfanyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
78754
Solid
1g
102830-49-9
MFCD00038253
n-boc-s-benzyl-d-cysteine, boc-d-cys bzl-oh, boc-s-benzyl-d-cysteine, n-tert-butyloxycarbonyl-s-benzylcysteine, 2s-3-benzylsulfanyl-2-tert-butoxycarbonyl amino propanoic acid, boc-s-benzyl-d-cys, boc-s-benzyl-l-cys, ambotzbaa5410, s-benzyl-n-tert-butoxycarbonyl-d-cysteine, alanine, 3-benzylthio-n-carboxy-, n-tert-butyl ester, l
CC(C)(C)OC(=O)NC(CSCC1=CC=CC=C1)C(=O)O
311.396
311.4
97%
Videos
SDS