missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-S-(4-methylbenzyl)-D-cysteine, 95%

Brand:  Alfa Aesar™ H62074.03

Additional Details : CAS Number : 61925-78-8 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15458658

  • 86.40 EUR / 1g


Estimated Shipment date
from Supplier 19-02-2019
Add to basket

Specifications

Chemical Identifiers

61925-78-8
325.423
CUNVVZWSABRKAL-CYBMUJFWSA-N
6992557
CC1=CC=C(C=C1)CSCC(C(=O)O)NC(=O)OC(C)(C)C
C16H23NO4S
MFCD00076924
boc-d-cys pmebzl-oh, boc-s-4-methylbenzyl-d-cysteine, boc-d-cys 4-mbzl-oh, boc-d-cys 4-mebzl-oh, boc-d-cys mbzl-oh, s-2-tert-butoxycarbonyl amino-3-4-methylbenzyl thio propanoic acid, 2s-2-tert-butoxy carbonyl amino-3-4-methylphenyl methyl sulfanyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-4-methylphenyl methyl sulfanyl propanoic acid, ambotzbaa5420, n-alpha-t-butyloxycarbonyl-s-4-methyl-benzyl-d-cysteine
(2S)-3-[(4-methylphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid

Specifications

N-Boc-S-(4-methylbenzyl)-D-cysteine
C16H23NO4S
boc-d-cys pmebzl-oh, boc-s-4-methylbenzyl-d-cysteine, boc-d-cys 4-mbzl-oh, boc-d-cys 4-mebzl-oh, boc-d-cys mbzl-oh, s-2-tert-butoxycarbonyl amino-3-4-methylbenzyl thio propanoic acid, 2s-2-tert-butoxy carbonyl amino-3-4-methylphenyl methyl sulfanyl propanoic acid, 2s-2-tert-butoxycarbonyl amino-3-4-methylphenyl methyl sulfanyl propanoic acid, ambotzbaa5420, n-alpha-t-butyloxycarbonyl-s-4-methyl-benzyl-d-cysteine
CC1=CC=C(C=C1)CSCC(C(=O)O)NC(=O)OC(C)(C)C
325.423
325.42
95%
61925-78-8
MFCD00076924
CUNVVZWSABRKAL-CYBMUJFWSA-N
(2S)-3-[(4-methylphenyl)methylsulfanyl]-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
6992557
Solid
1g
Videos
SDS